Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DQW1 Antibody (Genox 3.53), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB1002702UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002702UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002702UV Supplier Novus Biologicals Supplier No. NB1002702UV

Mouse Monoclonal Antibody

HLA DQW1 Monoclonal specifically detects HLA DQW1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen HLA DQW1
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone Genox 3.53
Conjugate DyLight 350
Dilution Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Gene Alias CELIAC1, DC1, HLA class II histocompatibility antigen, DQ beta 1 chain, HLA-DQB, IDDM1, LB-E12, MB1, MHC class II antigen DQB1, MHC class II antigen HLA-DQ-beta-1, MHC class II DQ beta chain, MHC class II HLA-DQ beta glycoprotein, MT1
Gene Symbols HLA-DQB1
Host Species Mouse
Immunogen Bristol 8 glycoprotein from the Bristol 8 B lymphoblastoid cell line.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Asthma, Diabetes Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3119
Test Specificity Human Leukocyte Antigen DQW1 (DC1 determinant of human HLA-DR antigens)
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.