Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-DR Antibody (L243) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10077855 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10077855 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10077855 Supplier Novus Biologicals Supplier No. NB10077855
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 29 publications

HLA-DR Monoclonal specifically detects HLA-DR in Baboon, Canine, Human, Primate samples. It is validated for Block/Neutralize, CyTOF, Dual RNAScope ISH-IHC, Electron Microscopy, ELISA, Flow Cytometry, Flow Cytometry (Cell Surface), Fluorophore-linked immunosorbent assay, Functional Assay, ICC/F, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In vitro Assay, Western Blot.

Specifications

Antigen HLA-DR
Applications Blocking Assay, CyTOF, Dual RNAScope ISH-IHC, Electron Microscopy, ELISA, Flow Cytometry, Flow Cytometry (Cell Surface), Fluorophore-Linked Immunosorbent Assay, Functional Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In vitro Assay, Western Blot
Classification Monoclonal
Clone L243
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Flow Cytometry < 0.5 ug/10^6 cells, ELISA reported in scientific literature (PMID 11070170), Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 7835916), Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:10 - 1:500, Functional reported in scientific literature (PMID 17900279), In vitro assay reported in scientific literature (PMID 19620786), Electron Microscopy reported in scientific literature (PMID 21660937), Flow (Cell Surface), CyTOF-ready, Block/Neutralize reported in scientific literature (PMID 12874328), Dual RNAscope ISH-IHC
Gene Accession No. P01903
Gene Alias FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW
Gene Symbols HLA-DRA
Host Species Mouse
Immunogen Human lymphoblastoid cell line (RPMI 8866).
Molecular Weight of Antigen 28 kDa
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cellular Markers, Diabetes Research, Immunology, Major Histocompatibility Complexes
Primary or Secondary Primary
Gene ID (Entrez) 3122
Test Specificity Human. Predicted cross-reactivity with Chimpanzee, Baboon, Cynomolgus, Rhesus, Marmoset, Tamarin, Squirrel Monkey and Canine.
Target Species Baboon, Canine, Human, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.