Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-DR Antibody (TAL 1B5), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB60989A750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB60989A750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB60989A750 Supplier Novus Biologicals Supplier No. NB600989AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA-DR Monoclonal specifically detects HLA-DR in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HLA-DR
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone TAL 1B5
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW
Gene Symbols HLA-DRA
Host Species Mouse
Immunogen Bristol 8 separated alpha chain preparation
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cellular Markers, Diabetes Research, Immunology, Major Histocompatibility Complexes
Primary or Secondary Primary
Gene ID (Entrez) 3122
Test Specificity Specific for Human Leukocyte Antigen DR, alpha subunit. May be used for HLA typing.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.