Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-DR Antibody (TAL 8.1), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N102708PEC5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N102708PEC5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N102708PEC5 Supplier Novus Biologicals Supplier No. NB1002708PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA-DR Monoclonal specifically detects HLA-DR in Human samples. It is validated for CyTOF, ELISA, Flow Cytometry, Immunohistochemistry, Radioimmune Assays (RIA), Western Blot.

Specifications

Antigen HLA-DR
Applications CyTOF, ELISA, Flow Cytometry, Immunohistochemistry, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone TAL 8.1
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW
Gene Symbols HLA-DRA
Host Species Mouse
Immunogen Cells from the B line WVB coated with mouse antiserum followed by WVB class II antigen.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cellular Markers, Diabetes Research, Immunology, Major Histocompatibility Complexes
Primary or Secondary Primary
Gene ID (Entrez) 3122.0
Test Specificity Specific for Human Leukocyte Antigens DR3, 6, 11, DRJ25. May be used for HLA typing.
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.