Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DR/DP Antibody (MEM-136), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N50447APCY7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N50447APCY7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N50447APCY7 Supplier Novus Biologicals Supplier No. NB500447APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA DR/DP Monoclonal specifically detects HLA DR/DP in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen HLA DR/DP
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-136
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW
Gene Symbols HLA-DRA
Host Species Mouse
Immunogen PHA-activated peripheral blood lymphocytes.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Cell Biology, Diabetes Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3122.0
Test Specificity The antibody MEM-136 recognizes common epitope on beta-chain of human HLA-DR and HLA-DP. It reacts with alpha/beta dimer as well as with dissociated beta-subunit. DR and DP are the isotypes of human MHC Class II molecules expressed on antigen-presenting cells (APC; dendritic cells, B lymphocytes, monocytes, macrophages).
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.