Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DRB1 Antibody (SPM289), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB006562 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB006562 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB006562 Supplier Novus Biologicals Supplier No. NBP234756AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA DRB1 Monoclonal specifically detects HLA DRB1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen HLA DRB1
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SPM289
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Clone P2-beta-3, DR1, DR-1, DR12, DR-12, DR13, DR-13, DR14, DR-14, DR16, DR-16, DR4, DR-4, DR5, DR-5, DR7, DR-7, DR8, DR-8, DR9, DR-9, DRB1, DRw10, DRw11, DRw8, DW2.2/DR2.2, FLJ75017, FLJ76359, HLA class II antigen beta chain, HLA class II histocompatibility antigen, DR-1 beta chain, HLA-DR1B, HLA-DRB, HLA-DRB1*, HLA-DRB2, HLA-DR-beta 1, human leucocyte antigen DRB1, leucocyte antigen DR beta 1 chain, leucocyte antigen DRB1, lymphocyte antigen DRB1, major histocompatibility complex, class II, DR beta 1, MHC class II antigen DRB1*1, MHC class II antigen DRB1*10, MHC class II antigen DRB1*11, MHC class II antigen DRB1*12, MHC class II antigen DRB1*13, MHC class II antigen DRB1*14, MHC class II antigen DRB1*15, MHC class II antigen DRB1*16, MHC class II antigen DRB1*3, MHC class II antigen DRB1*4, MHC class II antigen DRB1*7, MHC class II antigen DRB1*8, MHC class II antigen DRB1*9, MHC class II antigen HLA-DR13, MHC class II HLA-DR beta 1 chain, MHC class II HLA-DR-beta cell surface gly
Gene Symbols HLA-DRB1
Host Species Mouse
Immunogen Activated human peripheral blood mononuclear cells
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Asthma, Cell Biology, Diabetes Research, Immunology
Primary or Secondary Primary
Test Specificity This monoclonal antibody reacts with the beta-chain of HLA-DR antigen, a member of MHC class II molecules. It does not cross react with HLA-DP and HLA-DQ. The L243 antibody recognizes a different epitope than the SPM289 monoclonal antibody, and these antibodies do not cross-block binding to each others respective epitopes. HLA-DR is a heterodimeric cell surface glycoprotein comprised of a 36kDa alpha (heavy) chain and a 28kDa beta (light) chain. It is expressed on B-cells, activated T-cells, monocytes/macrophages, dendritic cells and other non-professional APCs. In conjunction with the CD3/TCR complex and CD4 molecules, HLA-DR is critical for efficient peptide presentation to CD4+ T cells. It is an excellent histiocytic marker in paraffin sections producing intense staining. True histiocytic neoplasms are similarly positive. HLA-DR antigens also occur on a variety of epithelial cells and their corresponding neoplastic counterparts.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.