Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DRB1 Antibody (TDR 31.1), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N250421P594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N250421P594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N250421P594 Supplier Novus Biologicals Supplier No. NBP250421PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA DRB1 Monoclonal specifically detects HLA DRB1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen HLA DRB1
Applications Flow Cytometry
Classification Monoclonal
Clone TDR 31.1
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Clone P2-beta-3, DR1, DR-1, DR12, DR-12, DR13, DR-13, DR14, DR-14, DR16, DR-16, DR4, DR-4, DR5, DR-5, DR7, DR-7, DR8, DR-8, DR9, DR-9, DRB1, DRw10, DRw11, DRw8, DW2.2/DR2.2, FLJ75017, FLJ76359, HLA class II antigen beta chain, HLA class II histocompatibility antigen, DR-1 beta chain, HLA-DR1B, HLA-DRB, HLA-DRB1*, HLA-DRB2, HLA-DR-beta 1, human leucocyte antigen DRB1, leucocyte antigen DR beta 1 chain, leucocyte antigen DRB1, lymphocyte antigen DRB1, major histocompatibility complex, class II, DR beta 1, MHC class II antigen DRB1*1, MHC class II antigen DRB1*10, MHC class II antigen DRB1*11, MHC class II antigen DRB1*12, MHC class II antigen DRB1*13, MHC class II antigen DRB1*14, MHC class II antigen DRB1*15, MHC class II antigen DRB1*16, MHC class II antigen DRB1*3, MHC class II antigen DRB1*4, MHC class II antigen DRB1*7, MHC class II antigen DRB1*8, MHC class II antigen DRB1*9, MHC class II antigen HLA-DR13, MHC class II HLA-DR beta 1 chain, MHC class II HLA-DR-beta cell surface gly
Gene Symbols HLA-DRB1
Host Species Mouse
Immunogen Ia fraction from Jy human B lymphoblastoid cell line. Class II from JY human B lymphoblastoid cell line.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Asthma, Cell Biology, Diabetes Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3123.0
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.