Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DRB3 Antibody (Bra30), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB389487 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB389487 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB389487 Supplier Novus Biologicals Supplier No. NBP308911JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA DRB3 Monoclonal antibody specifically detects HLA DRB3 in Human, Canine, Chimpanzee, Monkey, Baboon, Squirrel samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen HLA DRB3
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone Bra30
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias DR7, HLA class II histocompatibility antigen, DR beta 3 chain, HLA class II histocompatibility antigen, DRB1-7 beta chain, HLA-DR3B, HLA-DR52, HLA-DRB1, human leucocyte antigen DRB3, major histocompatibility complex, class II, DR beta 3, MGC117330, MHC class II antigen DR beta 3 chain, MHC class II antigen DRB3, MHC class II HLA-DR beta 3 chain
Host Species Mouse
Immunogen Human REH cells (non-T, non-B leukemia cell line)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Asthma, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3125
Target Species Human, Canine, Chimpanzee, Monkey, Baboon
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.