Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DRB3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP133666 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP133666 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP133666 Supplier Novus Biologicals Supplier No. NBP133666
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HLA DRB3 Polyclonal specifically detects HLA DRB3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HLA DRB3
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Alias DR7, HLA class II histocompatibility antigen, DR beta 3 chain, HLA class II histocompatibility antigen, DRB1-7 beta chain, HLA-DR3B, HLA-DR52, HLA-DRB1, human leucocyte antigen DRB3, major histocompatibility complex, class II, DR beta 3, MGC117330, MHC class II antigen DR beta 3 chain, MHC class II antigen DRB3, MHC class II HLA-DR beta 3 chain
Gene Symbols HLA-DRB3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human HLA-DRB3. The exact sequence is proprietary.
Molecular Weight of Antigen 30 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Asthma, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3125
Test Specificity Tested in human. Not yet tested in any other species.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.