Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DRB4 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB094826 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094826 100 μL
NB094827 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB094826 Supplier Novus Biologicals Supplier No. NBP303662100UL

Rabbit Polyclonal Antibody

HLA DRB4 Polyclonal specifically detects HLA DRB4 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen HLA DRB4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias class II histocompatibility antigen HLA DR alpha, beta1-0307, DR12, DR-12, DR13, DR-13, DR14, DR-14, DR4, DR-4, DRB1 transplantation antigen, DRB4, HLA class II histocompatibility antigen, DR beta 4 chain, HLA class II histocompatibility antigen, DRB1-12 beta chain, HLA class II histocompatibility antigen, DRB1-4 beta chain, HLA-DR4B, human leucocyte antigen DRB4, leucocyte antigen DRB1, leukocyte antigen, major histocompatibility complex, class II, DR beta 1, major histocompatibility complex, class II, DR beta 4, MHC class I antigen DRB1*12, MHC class I antigen DRB1*4, MHC class II antigen DRB1*12, MHC class II antigen DRB1*13, MHC class II antigen DRB1*14, MHC class II antigen DRB1*4, MHC class II antigen DRB4, MHC class II antigen HLA-DRB1, MHC class II antigen HLA-DR-beta, MHC class II HLA-DR-beta-7, MHC class2 antigen, MHC HLA DR-beta chain
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 30-227 of human HLA DRB4 (NP_068818.4).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Asthma, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3126
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.