Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-E Antibody (MEM-E/07), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N50310PECY5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N50310PECY5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N50310PECY5 Supplier Novus Biologicals Supplier No. NB500310PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA-E Monoclonal specifically detects HLA-E in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen HLA-E
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-E/07
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1
Gene Symbols HLA-E
Host Species Mouse
Immunogen Bacterially expressed recombinant HLA-E refolded with beta2-microglobulin and peptide.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Asthma, Diabetes Research, Immunology, Major Histocompatibility Complexes
Primary or Secondary Primary
Gene ID (Entrez) 3133.0
Test Specificity The antibody MEM-E/07 recognized native surface-expressed HLA-E, but not denaturated heavy chain of HLA-E. HLA-E belongs to the MHC Class I molecules (MHC Class Ib; nonclassical) and it is expressed on many types of the human cells. The published results revealed that the antibody cross-reacts with some classical MHC Class I molecules (MHC Class Ia): HLA-B7 (strongly), HLA-B8 (moderately), HLA-B27, -B44 (weakly).
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.