Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA G Antibody (2A12), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N144925P594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N144925P594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N144925P594 Supplier Novus Biologicals Supplier No. NBP144925PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA G Monoclonal specifically detects HLA G in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen HLA G
Applications Flow Cytometry
Classification Monoclonal
Clone 2A12
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias b2 microglobulin, HLA class I histocompatibility antigen, alpha chain G, HLA class I molecule, HLA G antigen, HLA-6.0, HLAG, HLA-G histocompatibility antigen, class I, G, major histocompatibility complex, class I, G, MHC class I antigen G, MHC-G, sHLA-G
Gene Symbols HLA-G
Host Species Mouse
Immunogen C-terminal amino acid sequence (22-mer) of soluble HLA-G5 and HLA-G6 proteins coupled to ovalbumin (NM_002127.5).
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Autoimmune Diseases, Cancer, Developmental Biology, Immunology, Inflammation, Innate Immunity, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 3135.0
Test Specificity The antibody 2A12 allows to discriminate between soluble HLA-G protein yielded by shedding from membrane-bound HLA-G forms that do not contain the intron 4-encoded epitope and soluble HLA-G5/HLA-G6 produced from intron 4-retaining alternatively spliced mRNAs. HLA-G belongs to the MHC Class I molecules (MHC Class Ib; nonclassical) and it is expressed on the surface of trophoblast cells.
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.