Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA G Antibody (87G) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP14312303 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP14312303 0.025 mg
NBP14312301 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP14312303 Supplier Novus Biologicals Supplier No. NBP1431230.025MG

Mouse Monoclonal Antibody has been used in 9 publications

HLA G Monoclonal specifically detects HLA G in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Flow (Cell Surface), CyTOF-ready, Block/Neutralize.

Specifications

Antigen HLA G
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Blocking Assay, CyTOF
Classification Monoclonal
Clone 87G
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Flow Cytometry 1 ug per million cells, ELISA, Immunocytochemistry/Immunofluorescence 1:100, Blocking/Neutralizing, CyTOF-ready
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. P17693
Gene Alias b2 microglobulin, HLA class I histocompatibility antigen, alpha chain G, HLA class I molecule, HLA G antigen, HLA-6.0, HLAG, HLA-G histocompatibility antigen, class I, G, major histocompatibility complex, class I, G, MHC class I antigen G, MHC-G
Gene Symbols HLA-G
Host Species Mouse
Immunogen 721.221 cells transfected with full length HLA G cDNA with high surface HLA G expression. [UniProt# P17693]
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3135
Test Specificity Membrane bound HLA G1 and soluble HLA G5.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.