Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA G Antibody (MEM-G/9), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB50314A594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB50314A594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB50314A594 Supplier Novus Biologicals Supplier No. NB500314AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA G Monoclonal specifically detects HLA G in Human, Mouse (Negative) samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen HLA G
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone MEM-G/9
Conjugate Alexa Fluor 594
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias b2 microglobulin, HLA class I histocompatibility antigen, alpha chain G, HLA class I molecule, HLA G antigen, HLA-6.0, HLAG, HLA-G histocompatibility antigen, class I, G, major histocompatibility complex, class I, G, MHC class I antigen G, MHC-G, sHLA-G
Gene Symbols HLA-G
Host Species Mouse
Immunogen Recombinant human HLA-G refolded with beta2-microglobulin and peptide (NM_002127.5)
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Autoimmune Diseases, Cancer, Developmental Biology, Immunology, Inflammation, Innate Immunity, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 3135
Test Specificity The antibody MEM-G/9 reacts with native form of human HLA-G1 on the cell surface as well as with soluble HLA-G5 isoform in its beta2-microglobulin associated form. Reactivity with HLA-G3 was also reported. The antibody MEM-G/9 is standard reagent thoroughly validated during 3rd International Conference on HLA-G (Paris, 2003).
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.