Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLM Antibody, Novus Biologicals™
SDP

Catalog No. NBP223625 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223625 0.1 mg
NB019363 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP223625 Supplier Novus Biologicals Supplier No. NBP223625
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HLM Polyclonal specifically detects HLM in Human samples. It is validated for Western Blot.

Specifications

Antigen HLM
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation PBS with 0.02% Sodium Azide
Gene Alias ORP4, ORP-4, OSBPL1, OSBPL4
Host Species Rabbit
Immunogen Reacts with residues 549-563 [EKGRWDEANTTEKQR] of the HLM protein.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23762
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.