Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLM Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
Antigen | HLM |
---|---|
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
HLM Polyclonal specifically detects HLM in Human samples. It is validated for Western Blot.Specifications
HLM | |
Western Blot | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
23762 | |
IgG | |
Affinity Purified |
Western Blot 1:500-1:1000 | |
Polyclonal | |
Rabbit | |
Human | |
ORP4, ORP-4, OSBPL1, OSBPL4 | |
Reacts with residues 549-563 [EKGRWDEANTTEKQR] of the HLM protein. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title