Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLTF Antibody, Novus Biologicals™
SDP

Catalog No. NB1001041 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001041 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001041 Supplier Novus Biologicals Supplier No. NB1001041
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

HLTF Polyclonal specifically detects HLTF in Human, Primate samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA, Western Blot.

Specifications

Antigen HLTF
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.2 - 1 ug/ml, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. Q14527
Gene Alias DNA-binding protein/plasminogen activator inhibitor 1 regulator, EC 3.6.1, EC 3.6.4.-, EC 6.3.2.-, helicase-like transcription factor, HIP116ARING finger protein 80, HIP116matrix associated, actin dependent regulator of chromatin, RNF80SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 3, subfamily a, member 3, Sucrose nonfermenting protein 2-like 3, ZBU1
Gene Symbols HLTF
Host Species Goat
Immunogen Peptide with sequence C-KQAKINEIRTLIDL corresponding to C-Terminus according to NP_003062.2, NP_620636.1.
Molecular Weight of Antigen 114 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 6596
Test Specificity NP_003062.2 and NP_620636.2 are varients that represent the same protein.
Target Species Human, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.