Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLTF Antibody, Novus Biologicals™
SDP

Catalog No. NBP183256 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP183256 0.1 mL
NB433082 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP183256 Supplier Novus Biologicals Supplier No. NBP183256
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

HLTF Polyclonal specifically detects HLTF in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen HLTF
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q14527
Gene Alias DNA-binding protein/plasminogen activator inhibitor 1 regulator, EC 3.6.1, EC 3.6.4.-, EC 6.3.2.-, helicase-like transcription factor, HIP116ARING finger protein 80, HIP116matrix associated, actin dependent regulator of chromatin, RNF80SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 3, subfamily a, member 3, Sucrose nonfermenting protein 2-like 3, ZBU1
Gene Symbols HLTF
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:LKKHGFKLGPAPKTLGFNLESGWGSGRAGPSYSMPVHAAVQMTTEQLKTEFDKLFEDLKEDDKTHEMEPAEAIETPLLPHQKQALAWMVSRENSKELPPFWEQRNDLYYNTITNFSEKDRPENVHGGILADDMGLGK
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Chromatin Research, Tumor Suppressors, Ubiquitin Proteasome Pathway, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 6596
Test Specificity Specificity of human HLTF antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.