Learn More
HM74A/PUMA-G/GPR109A/NIACR1 Antibody (245106), DyLight 350, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | HM74A/PUMA-G/GPR109A/NIACR1 |
| Applications | CyTOF, Flow Cytometry |
| Classification | Monoclonal |
| Clone | 245106 |
| Conjugate | DyLight 350 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | GPR109A, HCA2, hydroxycarboxylic acid receptor 2, NIACR1 |
| Host Species | Rat |
| Immunogen | NS0 mouse myeloma cell line transfected with human HM74A/GPR109A, Met1-Pro363, Accession # Q8TDS4 |
| Purification Method | Protein A or G purified from hybridoma culture supernatant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.