Learn More
HM74A/PUMA-G/GPR109A/NIACR1 Antibody (MM0329-6S24), DyLight 680, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | HM74A/PUMA-G/GPR109A/NIACR1 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | MM0329-6S24 |
| Conjugate | DyLight 680 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry |
| Gene Accession No. | Q8TDS4.1 |
| Gene Alias | GPR109A, HCA2, hydroxycarboxylic acid receptor 2, NIACR1 |
| Gene Symbols | HCAR2 |
| Host Species | Rat |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.