Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HM74A/PUMA-G/GPR109A/NIACR1 Antibody (MM0329-6S24), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N212358P594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N212358P594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N212358P594 Supplier Novus Biologicals Supplier No. NBP212358PEATT594
Add to Cart
Add to Cart

Rat Monoclonal Antibody

HM74A/PUMA-G/GPR109A/NIACR1 Monoclonal specifically detects HM74A/PUMA-G/GPR109A/NIACR1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen HM74A/PUMA-G/GPR109A/NIACR1
Applications Flow Cytometry
Classification Monoclonal
Clone MM0329-6S24
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias GPR109A, HCA2, hydroxycarboxylic acid receptor 2, NIACR1
Gene Symbols HCAR2
Host Species Rat
Immunogen Human recombinant HM74A/PUMA-G/GPR109A/NIACR1
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 338442.0
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.