Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMG-CoA Reductase/HMGCR Antibody (CL0259), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP26161625UL
Description
HMG-CoA Reductase/HMGCR Monoclonal specifically detects HMG-CoA Reductase/HMGCR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
HMG-CoA Reductase/HMGCR | |
Monoclonal | |
Unconjugated | |
PBS (pH 7.2), 40% glycerol with 0.02% Sodium Azide | |
HMGCR | |
This antibody was developed against a recombinant protein corresponding to amino acids: HIPAYKLETLMETHERGVSIRRQLLSKKLSEPSSLQYLPYRDYNYSLVMGACCENVIGYMPIPVGVAGPLCLDEKEFQVPMATTEGCLVA | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
IgG2a |
Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown | |
CL0259 | |
Western Blot 1 μg/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200, Simple Western (Negative), Knockdown Validated | |
3-hydroxy-3-methylglutaryl CoA reductase (NADPH), 3-hydroxy-3-methylglutaryl-CoA reductase, 3-hydroxy-3-methylglutaryl-Coenzyme A reductase, EC 1.1.1, EC 1.1.1.34, HMG-CoA reductase, hydroxymethylglutaryl-CoA reductase, LDLCQ3 | |
Mouse | |
Protein A purified | |
RUO | |
3156 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction