Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMG-CoA Reductase/HMGCR Antibody (CL0259), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $646.00
Specifications
Antigen | HMG-CoA Reductase/HMGCR |
---|---|
Clone | CL0259 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
HMG-CoA Reductase/HMGCR Monoclonal specifically detects HMG-CoA Reductase/HMGCR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
HMG-CoA Reductase/HMGCR | |
Monoclonal | |
Purified | |
Human | |
3156 | |
This antibody was developed against a recombinant protein corresponding to amino acids: HIPAYKLETLMETHERGVSIRRQLLSKKLSEPSSLQYLPYRDYNYSLVMGACCENVIGYMPIPVGVAGPLCLDEKEFQVPMATTEGCLVA | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
CL0259 | |
Unconjugated | |
Mouse | |
3-hydroxy-3-methylglutaryl CoA reductase (NADPH), 3-hydroxy-3-methylglutaryl-CoA reductase, 3-hydroxy-3-methylglutaryl-Coenzyme A reductase, EC 1.1.1, EC 1.1.1.34, HMG-CoA reductase, hydroxymethylglutaryl-CoA reductase, LDLCQ3 | |
HMGCR | |
IgG2a | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title