Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMG-Lysine Rabbit anti-All, Unconjugated, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$192.35 - $452.71
Specifications
Antigen | HMG-Lysine |
---|---|
Concentration | Please refer to lot specific datasheet. |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABS2108-25UL
|
MilliporeSigma
ABS210825UL |
25 μL |
Each for $192.35
|
|
|||||
ABS2108-100
|
MilliporeSigma
ABS2108100UL |
100 μL |
Each for $452.71
|
|
|||||
Description
Anti-HMG-Lysine, Cat. No. ABS2108, is a rabbit polyclonal antibody that detects hydroxymethylglutaryl-lysine conjugated proteins and has been tested for use in Western Blotting.Specifications
HMG-Lysine | |
Western Blot | |
Unconjugated | |
Rabbit | |
Signaling | |
Purified rabbit polyclonal antibody in PBS with 0.1% sodium azide. | |
3-hydroxy-3-methyl-glutaryl (HMG)-Lysine conjugated to BSA. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. | |
Antibodies |
Please refer to lot specific datasheet. | |
Polyclonal | |
Purified | |
RUO | |
All species | |
3-hydroxy-3-methyl-glutary-Lysine | |
IgG | |
Affinity Purified | |
This rabbit polyclonal antibody detects BSA-conjugated 3-hydroxy-3-methyl-glutaryl (HMG)-Lysine. It recognizes BSA chemically modified to contain HMG-lysine modifications, but not unmodified BSA. | |
∽75 kDa observed (BSA conjugate). Uncharacterized bands may be observed in some lysate(s). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title