Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMG20A Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB047205 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB047205 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB047205 Supplier Novus Biologicals Supplier No. NBP297735B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HMG20A Polyclonal specifically detects HMG20A in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HMG20A
Applications Immunofluorescence
Classification Polyclonal
Conjugate Biotin
Dilution Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias FLJ10739, high-mobility group 20A, HMG box domain containing 1, HMG box-containing protein 20A, HMG domain-containing protein 1, HMGX1HMG domain-containing protein HMGX1, HMGXB1high mobility group protein 20A
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human HMG20A fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10363
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.