Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMG20A Antibody (OTI2H4), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP271596R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271596R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271596R Supplier Novus Biologicals Supplier No. NBP271596R

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

HMG20A Monoclonal specifically detects HMG20A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen HMG20A
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2H4
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ10739, high-mobility group 20A, HMG box domain containing 1, HMG box-containing protein 20A, HMG domain-containing protein 1, HMGX1HMG domain-containing protein HMGX1, HMGXB1high mobility group protein 20A
Gene Symbols HMG20A
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-281 of human HMG20A (NP_060670) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Chromatin Research, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10363.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.