Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMG2L1 Antibody (OTI1H10), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB288806 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288806 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288806 Supplier Novus Biologicals Supplier No. NBP271538APC

Mouse Monoclonal Antibody

HMG2L1 Monoclonal antibody specifically detects HMG2L1 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen HMG2L1
Applications Western Blot
Classification Monoclonal
Clone OTI1H10
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias High mobility group protein 2-like 1, high-mobility group protein 2-like 1, HMG box domain containing 4, HMG domain-containing protein 4, HMG2L1HMG box-containing protein 4, HMGBCG, Protein HMGBCG, THC211630
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-348 of human HMGXB4 (NP_005478) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10042
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.