Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMGB1/HMG-1 Antibody, Novus Biologicals™
SDP

Catalog No. NB1002322 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002322 0.1 mL
NB1002322SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1002322 Supplier Novus Biologicals Supplier No. NB1002322
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 22 publications

HMGB1/HMG-1 Polyclonal specifically detects HMGB1/HMG-1 in Bovine, Canine, Human, Mouse, Rabbit, Rat, Sheep samples. It is validated for ELISA, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown, Simple Western, Western Blot.

Specifications

Antigen HMGB1/HMG-1
Applications ELISA, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown, Simple Western, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1.0 ug/ml, Simple Western 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:100-1:250, Immunocytochemistry/Immunofluorescence 0.05 ug/ml, Immunohistochemistry-Paraffin 1:100-1:250, Flow (Intracellular)
Formulation Tris-Glycine with 0.02% Sodium Azide
Gene Accession No. P09429
Gene Alias Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein
Gene Symbols HMGB1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human HMGB1 protein sequence (between residues 100-200). [UniProt #P09429]
Molecular Weight of Antigen 29 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Chromatin Research, Histones and Modified Histones, Neuroscience, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 3146
Test Specificity Human, mouse, rat, dog, bovine, sheep. Predicted to react with porcine, equine and rabbit based on 100% sequence homology.
Target Species Bovine, Mouse, Canine, Human, Mouse, Rabbit, Rabbit, Rat, Sheep
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.