Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMGB1/HMG-1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB355099 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355099 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355099 Supplier Novus Biologicals Supplier No. NB1002322PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HMGB1/HMG-1 Polyclonal specifically detects HMGB1/HMG-1 in Bovine, Canine, Human, Mouse, Rabbit, Rat, Sheep samples. It is validated for ELISA, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown, Western Blot.

Specifications

Antigen HMGB1/HMG-1
Applications ELISA, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown, Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), Knockdown Validated
Formulation PBS
Gene Alias Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human HMGB1 protein sequence (between residues 100-200). [UniProt #P09429]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Chromatin Research, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3146
Target Species Bovine, Canine, Human, Mouse, Rabbit, Rat, Sheep
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.