Learn More
Invitrogen™ HMGB1 Polyclonal Antibody, DyLight™ 488

Description
HMGB1 Polyclonal Antibody for ELISA, Flow, ICC/IF, IHC (P), Western Blot
Specifications
Specifications
| Antigen | HMGB1 |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.81 mg/mL |
| Conjugate | DyLight 488 |
| Formulation | 50mM sodium borate with 0.05% sodium azide |
| Gene | Hmgb1 |
| Gene Accession No. | P09429, P10103, P63158, P63159, Q6YKA4 |
| Gene Alias | Ac2-008; amphoterin; Amphoterin antibody; DEF; DKFZp686A04236; Heparin-binding protein p30; high mobility group 1; high mobility group 1 protein; high mobility group box 1; high mobility group protein 1; High mobility group protein B1; high mobility group protein HMG1; high-mobility group (nonhistone chromosomal) protein 1; high-mobility group box 1; high-mobility-group protein; Hmg1; HMG-1; HMG3; HMG3 antibody; hmgb 1; HMGB1; hmgb-1; HMGB1 protein; hypothetical protein; non-histone protein HMG1; p30; RCJMB04_15a21; RP11-550P23.1; SBP-1; Sulfoglucuronyl carbohydrate binding protein; unnamed protein product |
| Gene Symbols | Hmgb1, LOC101112071 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.