Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMGB3/HMG4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224512 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224512 0.1 mg
NBP224512SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224512 Supplier Novus Biologicals Supplier No. NBP224512
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HMGB3/HMG4 Polyclonal specifically detects HMGB3/HMG4 in Human, Mouse, Bovine, Canine, Chicken, Equine, Primate samples. It is validated for Western Blot.

Specifications

Antigen HMGB3/HMG4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1.0 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. O15347
Gene Alias High mobility group protein 2a, High mobility group protein 4, high mobility group protein B3, high-mobility group (nonhistone chromosomal) protein 4, high-mobility group box 3, HMG-2a, HMG2AHMG-4, HMG4MGC90319, non-histone chromosomal protein
Gene Symbols HMGB3
Host Species Rabbit
Immunogen A portion of amino acids 150-200 of human HMGB3 was used as the immunogen.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Chromatin Research, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 3149
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_c).
Target Species Human, Mouse, Bovine, Canine, Chicken, Equine, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.