Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMGB3/HMG4 Rabbit anti-Human, DyLight 650, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB106535 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB106535 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB106535 Supplier Novus Biologicals Supplier No. NBP298887C

Rabbit Polyclonal Antibody

HMGB3/HMG4 Polyclonal specifically detects HMGB3/HMG4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen HMGB3/HMG4
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Formulation 50 mM Sodium Borate
Gene Alias High mobility group protein 2a, High mobility group protein 4, high mobility group protein B3, high-mobility group (nonhistone chromosomal) protein 4, high-mobility group box 3, HMG-2a, HMG2AHMG-4, HMG4MGC90319, non-histone chromosomal protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human HMGB3/HMG4.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 3149
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.