Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMGCS1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP236554 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP236554 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP236554 Supplier Novus Biologicals Supplier No. NBP236554
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HMGCS1 Polyclonal specifically detects HMGCS1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen HMGCS1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_001091742.1
Gene Alias 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) synthase, 3-hydroxy-3-methylglutaryl coenzyme A synthase, 3-hydroxy-3-methylglutaryl-CoA synthase 1 (soluble), 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1 (soluble), EC 2.3.3.10, HMG-CoA synthase, HMGCS, hydroxymethylglutaryl-CoA synthase, cytoplasmic, MGC90332
Gene Symbols HMGCS1
Host Species Rabbit
Immunogen The immunogen recognized by the antibody maps to a region between residue 470 to 520 of human 3-hydroxy-3-methylglutaryl-CoA Synthase 1 using the numbering given in entry NP_001091742.1.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 3157
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.