Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMGN1/HMG14 Antibody, Novus Biologicals™
SDP

Catalog No. NBP214842SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP214842SS 0.025 mg
NBP214842 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP214842SS Supplier Novus Biologicals Supplier No. NBP214842SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HMGN1/HMG14 Polyclonal specifically detects HMGN1/HMG14 in Human, Porcine, Bovine, Canine, Primate, Rabbit samples. It is validated for Western Blot.

Specifications

Antigen HMGN1/HMG14
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml
Formulation PBS containing 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. NP_004956.5
Gene Alias FLJ31471, High mobility group nucleosome-binding domain-containing protein 1, high-mobility group (nonhistone chromosomal) protein 14, high-mobility group nucleosome binding 1, high-mobility group nucleosome binding domain 1, HMG14FLJ27265, MGC104230, MGC117425, non-histone chromosomal protein HMG-14, nonhistone chromosomal protein HMG-14
Gene Symbols HMGN1
Host Species Rabbit
Immunogen A portion of amino acids 1-50 from human HMG14 was used as the immunogen for this antibody.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Histone Deacetylases, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3150
Test Specificity Reacts with Cow, Dog, Human, New World monkey, Pig, Rabbit, Rhesus Monkey and White-cheeked gibbon. Predicted to react with Mouse, Elephant, Chimpanzee, Chicken, Guinea Pig, Mole Rat, Opossum and Carolina anole. The HMG14 amino acid sequence used as immunogen for this antibody is 100% homologous in human, cow, new world monkey, dog, rhesus monkey, white-cheeked gibbon, rabbit, and pig, 94% in guinea pig, mole rat, elephant, chimpanzee, and orangutan, 83% in panda, opossum, mouse, and rat, 78% in zebra finch, 72% in Carolina anole, and wild turkey, and 61% homologouus in platypus.
Target Species Human, Pig, Bovine, Canine, Primate, Rabbit
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.