Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMGN1/HMG14 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$149.33 - $394.26
Specifications
| Antigen | HMGN1/HMG14 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 2 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP214842SS
![]() |
Novus Biologicals
NBP214842SS |
0.025 mg |
Each for $149.33
|
|
|||||
NBP214842
![]() |
Novus Biologicals
NBP214842 |
0.1 mg |
Each for $394.26
|
|
|||||
Description
HMGN1/HMG14 Polyclonal specifically detects HMGN1/HMG14 in Human, Porcine, Bovine, Canine, Primate, Rabbit samples. It is validated for Western Blot.Specifications
| HMGN1/HMG14 | |
| Western Blot 2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Pig, Bovine, Canine, Primate, Rabbit | |
| NP_004956.5 | |
| 3150 | |
| A portion of amino acids 1-50 from human HMG14 was used as the immunogen for this antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Histone Deacetylases, Stem Cell Markers | |
| PBS containing 0.05% BSA with 0.05% Sodium Azide | |
| FLJ31471, High mobility group nucleosome-binding domain-containing protein 1, high-mobility group (nonhistone chromosomal) protein 14, high-mobility group nucleosome binding 1, high-mobility group nucleosome binding domain 1, HMG14FLJ27265, MGC104230, MGC117425, non-histone chromosomal protein HMG-14, nonhistone chromosomal protein HMG-14 | |
| HMGN1 | |
| IgG | |
| Protein A purified | |
| Reacts with Cow, Dog, Human, New World monkey, Pig, Rabbit, Rhesus Monkey and White-cheeked gibbon. Predicted to react with Mouse, Elephant, Chimpanzee, Chicken, Guinea Pig, Mole Rat, Opossum and Carolina anole. The HMG14 amino acid sequence used as immunogen for this antibody is 100% homologous in human, cow, new world monkey, dog, rhesus monkey, white-cheeked gibbon, rabbit, and pig, 94% in guinea pig, mole rat, elephant, chimpanzee, and orangutan, 83% in panda, opossum, mouse, and rat, 78% in zebra finch, 72% in Carolina anole, and wild turkey, and 61% homologouus in platypus. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title