Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMGN1/HMG14 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB094698 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094698 100 μL
NB094699 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB094698 Supplier Novus Biologicals Supplier No. NBP303219100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HMGN1/HMG14 Polyclonal specifically detects HMGN1/HMG14 in Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen HMGN1/HMG14
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias FLJ31471, High mobility group nucleosome-binding domain-containing protein 1, high-mobility group (nonhistone chromosomal) protein 14, high-mobility group nucleosome binding 1, high-mobility group nucleosome binding domain 1, HMG14FLJ27265, MGC104230, MGC117425, non-histone chromosomal protein HMG-14, nonhistone chromosomal protein HMG-14
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human HMGN1/HMG14 (NP_004956.5).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3150
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.