Learn More
Description
Specifically detects HMW E. coli Host Cell Proteins in E. coli samples, and it is validated for ELISA, Western Blotting
Specifications
Specifications
| Antigen | HMW E.Coli Host Cell Proteins |
| Applications | Western Blot, ELISA, Dot Blot |
| Classification | Polyclonal |
| Concentration | 77.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:1000, ELISA 1:1000-1:2000, Dot Blot |
| Formulation | 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide |
| Host Species | Rabbit |
| Immunogen | HMW E.Coli Host Cell Proteins Antibody was prepared by repeated immunizations with a mixture high molecular weight (HMW) proteins from E.Coli (BL21). |
| Purification Method | Delipidation and Defibrination |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
