Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HN1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP132510 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP132510 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP132510 Supplier Novus Biologicals Supplier No. NBP132510
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HN1 Polyclonal specifically detects HN1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen HN1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.91mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, ELISA 1:1000-1:10000, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS, 20% Glycerol (pH7) with 0.025% Proclin 300
Gene Alias Androgen-regulated protein 2, ARM2hematological and neurological expressed 1 protein, hematological and neurological expressed 1, HN1A, OPMRX60
Gene Symbols JPT1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human HN1. The exact sequence is proprietary.
Molecular Weight of Antigen 16 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 51155
Test Specificity Tested in human. Predicted cross-reactivity based on sequence identity: Mouse (88%), Rat (82%), Cow (88%), Dog (83%), and Monkey (100%). Not yet tested in any other species.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.