Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HNF-3 alpha/FoxA1 Antibody (3B2), Novus Biologicals™
SDP

Catalog No. NB297833 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297833 100 μL
NB297832 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297833 Supplier Novus Biologicals Supplier No. NBP326572100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

HNF-3 alpha/FoxA1 Monoclonal antibody specifically detects HNF-3 alpha/FoxA1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation

Specifications

Antigen HNF-3 alpha/FoxA1
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 3B2
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias forkhead box A1, Forkhead box protein A1, hepatocyte nuclear factor 3, alpha, hepatocyte nuclear factor 3-alpha, HNF-3A, HNF-3-alpha, HNF3ATCF-3A, MGC33105, TCF3A, Transcription factor 3A
Host Species Rabbit
Immunogen A synthesized peptide derived from Human HNF-3 alpha/FoxA1 [UniProt P55317]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3169
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.