Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HNF-3 beta/FoxA2 Antibody (OTI3C10), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP270904IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270904IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270904IR Supplier Novus Biologicals Supplier No. NBP270904IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HNF-3 beta/FoxA2 Monoclonal specifically detects HNF-3 beta/FoxA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HNF-3 beta/FoxA2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI3C10
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias forkhead box A2, Forkhead box protein A2, hepatic nuclear factor-3-beta, hepatocyte nuclear factor 3, beta, hepatocyte nuclear factor 3-beta, HNF-3B, HNF-3-beta, HNF3BTCF-3B, MGC19807, TCF3B, Transcription factor 3B
Gene Symbols FOXA2
Host Species Mouse
Immunogen Recombinant protein expressed in E.coli corresponding to amino acids 300-458 of human HNF-3 beta/FoxA2.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3170.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.