Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HNF-4 alpha/NR2A1 Antibody (843716), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB303512 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303512 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303512 Supplier Novus Biologicals Supplier No. FAB4605S

Mouse Monoclonal Antibody

HNF-4 alpha/NR2A1 Monoclonal antibody specifically detects HNF-4 alpha/NR2A1 in Human samples. It is validated for Western Blot, Western Blot, Flow Cytometry, Immunocytochemistry

Specifications

Antigen HNF-4 alpha/NR2A1
Applications Western Blot, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 843716
Conjugate Alexa Fluor 750
Dilution Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias FLJ39654, hepatic nuclear factor 4 alpha, hepatocyte nuclear factor 4, alpha, hepatocyte nuclear factor 4-alpha, HNF4a7, HNF-4-alpha, HNF4alpha10/11/12, HNF4HNF4a8, MODY, MODY1, NR2A1HNF4a9, NR2A21, Nuclear receptor subfamily 2 group A member 1, TCF, TCF-14, TCF14HNF4alpha, Transcription factor 14, Transcription factor HNF-4, transcription factor-14
Host Species Mouse
Immunogen E. coli-derived recombinant human HNF-4 alpha/NR2A1, Val130-Ser330, Accession # P41235
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3172
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.