Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HNF-4 alpha/NR2A1 Antibody (SN72-03), Novus Biologicals™
SDP

Catalog No. NBP267679 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267679 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267679 Supplier Novus Biologicals Supplier No. NBP267679

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Monoclonal Antibody

HNF-4 alpha/NR2A1 Monoclonal specifically detects HNF-4 alpha/NR2A1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen HNF-4alpha/NR2A1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SN72-03
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:50-:100, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1:500
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias FLJ39654, hepatic nuclear factor 4 alpha, hepatocyte nuclear factor 4, alpha, hepatocyte nuclear factor 4-alpha, HNF4a7, HNF-4-alpha, HNF4alpha10/11/12, HNF4HNF4a8, MODY, MODY1, NR2A1HNF4a9, NR2A21, Nuclear receptor subfamily 2 group A member 1, TCF, TCF-14, TCF14HNF4alpha, Transcription factor 14, Transcription factor HNF-4, transcription factor-14
Gene Symbols HNF4A
Host Species Rabbit
Immunogen Synthetic peptide within Human HNF-4 alpha/NR2A1 aa 21-70/ 474. (SwissProt: P41235 Human)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3172
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.