Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HNF-6/ONECUT1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB326104 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB326104 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB326104 Supplier Novus Biologicals Supplier No. NBP299314MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HNF-6/ONECUT1 Polyclonal antibody specifically detects HNF-6/ONECUT1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen HNF-6/ONECUT1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias HNF6Ahepatocyte nuclear factor 6, alpha, HNF6hepatocyte nuclear factor 6, HNF-6One cut domain family member 1, one cut homeobox 1one cut domain, family member 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human HNF-6/ONECUT1.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3175
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.