Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HNF1 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB289987 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289987 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289987 Supplier Novus Biologicals Supplier No. NBP232126PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HNF1 Polyclonal antibody specifically detects HNF1 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot (Negative)

Specifications

Antigen HNF1
Applications Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate PE
Dilution Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative)
Formulation PBS
Gene Alias HNF1 homeobox A, IDDM20, LFB1transcription factor 1, hepatic, LF-B1, hepatic nuclear factor (HNF1), albuminproximal factor, MODY3, TCF1hepatic, Transcription factor 1
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 581 to 631 of human HNF1 Homeobox A using the numbering given in entry NP_000536.5 (GeneID 6927).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6927
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.