Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP A1 Antibody (4B10), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB100672X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100672X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100672X Supplier Novus Biologicals Supplier No. NB100672AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

hnRNP A1 Monoclonal specifically detects hnRNP A1 in Bovine, Canine, Human, Mouse samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunoprecipitation, KnockDown, Microarray, Western Blot.

Specifications

Antigen hnRNP A1
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, KnockDown, Microarray, Western Blot
Classification Monoclonal
Clone 4B10
Conjugate Alexa Fluor 488
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockdown Validated
Gene Accession No. P09651
Gene Alias Helix-destabilizing protein, heterogeneous nuclear ribonucleoprotein A1, heterogeneous nuclear ribonucleoprotein A1B protein, heterogeneous nuclear ribonucleoprotein B2 protein, heterogeneous nuclear ribonucleoprotein core protein A1, hnRNP A1, hnRNP core protein A1, hnRNPA1, hnRNP-A1, HNRPA1MGC102835, nuclear ribonucleoprotein particle A1 protein, single-strand DNA-binding protein UP1, Single-strand RNA-binding protein
Gene Symbols HNRNPA1
Host Species Mouse
Immunogen Full length partially purified human hnRNP A1. [UniProt# P09651]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3178
Test Specificity Does not react with hnRNP A2 or B
Target Species Bovine, Canine, Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.