Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP A1 Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB048252 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB048252 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB048252 Supplier Novus Biologicals Supplier No. NBP297829V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

hnRNP A1 Polyclonal specifically detects hnRNP A1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen hnRNP A1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Helix-destabilizing protein, heterogeneous nuclear ribonucleoprotein A1, heterogeneous nuclear ribonucleoprotein A1B protein, heterogeneous nuclear ribonucleoprotein B2 protein, heterogeneous nuclear ribonucleoprotein core protein A1, hnRNP A1, hnRNP core protein A1, hnRNPA1, hnRNP-A1, HNRPA1MGC102835, nuclear ribonucleoprotein particle A1 protein, single-strand DNA-binding protein UP1, Single-strand RNA-binding protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human hnRNP A1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3178
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.