Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP A2B1 Antibody (DP3B3), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. N1206102A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1206102A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1206102A30 Supplier Novus Biologicals Supplier No. NB1206102AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

hnRNP A2B1 Monoclonal specifically detects hnRNP A2B1 in Human, Mouse, Rat, Bovine, Canine, Chicken, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated, Knockdown Validated.

Specifications

Antigen hnRNP A2B1
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoassay, KnockDown
Classification Monoclonal
Clone DP3B3
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated, Knockdown Validated
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DKFZp779B0244, FLJ22720, heterogeneous nuclear ribonucleoprotein A2/B1, heterogeneous nuclear ribonucleoprotein B1, heterogeneous nuclear ribonucleoproteins A2/B1, hnRNP A2/ hnRNP B1, HNRNPA2, HNRNPB1, HNRPA2, HNRPA2B1hnRNP A2/B1, HNRPB1, nuclear ribonucleoprotein particle A2 protein, RNPA2, SNRPB1
Gene Symbols HNRNPA2B1
Host Species Mouse
Immunogen Bacterially expressed human hnRNP A2 fusion protein.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3181
Target Species Human, Mouse, Rat, Bovine, Canine, Chicken, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.