Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP A2B1 Antibody (HL1706) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB295109 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295109 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295109 Supplier Novus Biologicals Supplier No. NBP325508
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

hnRNP A2B1 Monoclonal antibody specifically detects hnRNP A2B1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen hnRNP A2B1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HL1706
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias DKFZp779B0244, FLJ22720, heterogeneous nuclear ribonucleoprotein A2/B1, heterogeneous nuclear ribonucleoprotein B1, heterogeneous nuclear ribonucleoproteins A2/B1, hnRNP A2/ hnRNP B1, HNRNPA2, HNRNPB1, HNRPA2, HNRPA2B1hnRNP A2/B1, HNRPB1, nuclear ribonucleoprotein particle A2 protein, RNPA2, SNRPB1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human hnRNP A2B1. The exact sequence is proprietary.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 3181
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.