Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP A2B1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP256497 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP256497 100 μL
NB403680 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP256497 Supplier Novus Biologicals Supplier No. NBP256497
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

hnRNP A2B1 Polyclonal specifically detects hnRNP A2B1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated.

Specifications

Antigen hnRNP A2B1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Knockout Validated, KnockDown Validated
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias DKFZp779B0244, FLJ22720, heterogeneous nuclear ribonucleoprotein A2/B1, heterogeneous nuclear ribonucleoprotein B1, heterogeneous nuclear ribonucleoproteins A2/B1, hnRNP A2 hnRNP B1, HNRNPA2, HNRNPB1, HNRPA2, HNRPA2B1hnRNP A2/B1, HNRPB1, nuclear ribonucleoprotein particle A2 protein, RNPA2, SNRPB1
Gene Symbols HNRNPA2B1
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RQEMQEVQSSRSGRGGNFGFGDSRG
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 3181
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.