Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hnRNP A2B1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | hnRNP A2B1 |
---|---|
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
hnRNP A2B1 Polyclonal specifically detects hnRNP A2B1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated.Specifications
hnRNP A2B1 | |
Polyclonal | |
Rabbit | |
Cancer, DNA Repair, DNA replication Transcription Translation and Splicing | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
3181 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RQEMQEVQSSRSGRGGNFGFGDSRG | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
DKFZp779B0244, FLJ22720, heterogeneous nuclear ribonucleoprotein A2/B1, heterogeneous nuclear ribonucleoprotein B1, heterogeneous nuclear ribonucleoproteins A2/B1, hnRNP A2 hnRNP B1, HNRNPA2, HNRNPB1, HNRPA2, HNRPA2B1hnRNP A2/B1, HNRPB1, nuclear ribonucleoprotein particle A2 protein, RNPA2, SNRPB1 | |
HNRNPA2B1 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title